Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031020.2 | NP_112282.2 | 9 - 10 | 1071 | 140 | |
GenBank mRNA | AB000783.1 | - | 9 - 10 | 1070 | 140 | |
AF346293.1 | - | 9 - 10 | 768 | 140 | ||
BC092193.1 | - | 9 - 10 | 1071 | 7313217 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |