Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770464.1 | XP_008768686.1 | 1 - 2 | 253 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |