Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001509.1 | NP_001001509.1 | 5 - 6 | 685 | 91 | |
GenBank mRNA | AY603424.1 | - | 5 - 6 | 685 | 91 | |
BC089874.1 | - | 5 - 6 | 689 | 7325453 | 91 | |
FQ210765.1 | - | 5 - 6 | 745 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |