Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipcef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170799.1 | NP_001164270.1 | 8 - 9 | 1156 | 74 | |
GenBank mRNA | AJ536192.1 | - | 8 - 9 | 1156 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |