Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053991.1 | NP_446443.1 | 1 - 2 | 140 | 66 | |
XM_006227827.2 | XP_006227889.1 | 2 - 3 | 585 | 66 | ||
XM_006227828.2 | XP_006227890.1 | 2 - 3 | 588 | 66 | ||
GenBank mRNA | AB089809.1 | - | 1 - 2 | 157 | 66 | |
BC158798.1 | - | 1 - 2 | 140 | 7316540 | 66 | |
CB583853.1 | - | 1 - 2 | 154 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |