Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107433.1 | NP_001100903.1 | 2 - 3 | 1258 | 109 | |
XM_006255674.2 | XP_006255736.1 | 4 - 5 | 1779 | 109 | ||
XM_006255675.2 | XP_006255737.1 | 4 - 5 | 1593 | 109 | ||
XM_008772515.1 | XP_008770737.1 | 3 - 4 | 1808 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |