Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022962.1 | NP_075251.1 | 6 - 7 | 1494 | 79 | |
GenBank mRNA | AF081144.1 | - | 5 - 6 | 1479 | 79 | |
AF081145.1 | - | 5 - 6 | 1479 | 79 | ||
AF081146.1 | - | 6 - 7 | 1494 | 79 | ||
AF081147.1 | - | 6 - 7 | 1494 | 79 | ||
AF111099.1 | - | 7 - 8 | 1820 | 79 | ||
U72487.1 | - | 5 - 6 | 1296 | 79 | ||
U78105.1 | - | 5 - 6 | 1621 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |