Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017175.2 | NP_058871.2 | 15 - 16 | 2160 | 120 | |
XM_006255245.2 | XP_006255307.1 | 15 - 16 | 3200 | 120 | ||
GenBank mRNA | BC061836.1 | - | 15 - 16 | 2061 | 5599989 | 120 |
D26180.1 | - | 15 - 16 | 2111 | 120 | ||
L35634.1 | - | 15 - 16 | 2003 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |