Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053988.1 | NP_446440.1 | 9 - 10 | 684 | 77 | |
XM_008772466.1 | XP_008770688.1 | 10 - 11 | 863 | 77 | ||
GenBank mRNA | BC087603.1 | - | 9 - 10 | 654 | 7312595 | 77 |
X66974.1 | - | 9 - 10 | 684 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |