Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107424.1 | NP_001100894.1 | 5 - 6 | 1062 | 118 | |
XM_006255498.2 | XP_006255560.1 | 6 - 7 | 1127 | 118 | ||
XM_006255499.2 | XP_006255561.1 | 6 - 7 | 1157 | 118 | ||
XM_006255500.2 | XP_006255562.1 | 6 - 7 | 1275 | 118 | ||
XM_008772500.1 | XP_008770722.1 | 6 - 7 | 1161 | 118 | ||
XM_008772501.1 | XP_008770723.1 | 6 - 7 | 1342 | 118 | ||
XM_008772502.1 | XP_008770724.1 | 6 - 7 | 1391 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |