Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728582.4 | XP_002728628.1 | 4 - 5 | 423 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |