Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013061.1 | NP_001013079.1 | 4 - 5 | 397 | 114 | |
GenBank mRNA | FQ218973.1 | - | 424 | 114 | ||
FQ228571.1 | - | 4 - 5 | 415 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |