Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_578474.1 | XP_578474.1 | 6 - 7 | 1004 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |