Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190236.1 | NP_001177165.1 | 10 - 11 | 1086 | 85 | |
NM_001190237.1 | NP_001177166.1 | 10 - 11 | 1083 | 85 | ||
NM_001190238.1 | NP_001177167.1 | 9 - 10 | 1074 | 85 | ||
NM_013013.2 | NP_037145.2 | 9 - 10 | 1077 | 85 | ||
XM_006256403.1 | XP_006256465.1 | 10 - 11 | 1095 | 85 | ||
GenBank mRNA | BC061759.1 | - | 9 - 10 | 1008 | 5600088 | 85 |
CB717025.1 | - | 5 - 6 | 434 | 85 | ||
FQ217536.1 | - | 9 - 10 | 1028 | 85 | ||
M19936.1 | - | 9 - 10 | 1027 | 85 | ||
S81353.1 | - | 9 - 10 | 1027 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |