Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773008.1 | XP_008771230.1 | 4 - 5 | 813 | 96 | |
XM_008773009.1 | XP_008771231.1 | 4 - 5 | 813 | 96 | ||
XM_008773010.1 | XP_008771232.1 | 4 - 5 | 813 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |