Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271106.1 | NP_001258035.1 | 1 - 2 | 213 | 81 | |
GenBank mRNA | AJ535460.1 | - | 1 - 2 | 189 | 81 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Ion Channel Rattus norvegicus Probe spans exons |