Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115032.1 | NP_001108504.1 | 3 - 4 | 450 | 62 | |
GenBank mRNA | BC158654.1 | - | 3 - 4 | 450 | 8368493 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |