Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076553.2 | NP_001070021.1 | 2 - 3 | 200 | 124 | |
NM_001271158.1 | NP_001258087.1 | 2 - 3 | 140 | 124 | ||
NM_012627.3 | NP_036759.2 | 2 - 3 | 200 | 124 | ||
XM_006256511.2 | XP_006256573.1 | 2 - 3 | 819 | 124 | ||
XM_006256512.2 | XP_006256574.1 | 3 - 4 | 322 | 124 | ||
XM_006256513.2 | XP_006256575.1 | 2 - 3 | 177 | 124 | ||
XM_006256515.2 | XP_006256577.1 | 2 - 3 | 742 | 124 | ||
XM_008772976.1 | XP_008771198.1 | 3 - 4 | 459 | 124 | ||
XM_008772978.1 | XP_008771200.1 | 2 - 3 | 909 | 124 | ||
GenBank mRNA | AF413572.1 | - | 2 - 3 | 183 | 123 | |
BC078880.1 | - | 2 - 3 | 140 | 7111554 | 124 | |
BG666532.1 | - | 2 - 3 | 229 | 124 | ||
BG668634.1 | - | 267 | 124 | |||
FQ231722.1 | - | 2 - 3 | 196 | 124 | ||
M64092.1 | - | 2 - 3 | 157 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |