Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008556.1 | NP_001008556.1 | 2 - 3 | 128 | 71 | |
GenBank mRNA | AY823319.1 | - | 2 - 3 | 128 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |