Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108407.1 | NP_001101877.1 | 5 - 6 | 450 | 65 | |
XM_006253674.2 | XP_006253736.1 | 5 - 6 | 548 | 65 | ||
XM_008771499.1 | XP_008769721.1 | 5 - 6 | 1504 | 65 | ||
GenBank mRNA | FQ218844.1 | - | 5 - 6 | 542 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |