Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012052.1 | NP_001012052.1 | 6 - 7 | 2200 | 116 | |
XM_006253808.2 | XP_006253870.1 | 6 - 7 | 2187 | 116 | ||
XM_006253809.2 | XP_006253871.1 | 5 - 6 | 1878 | 116 | ||
GenBank mRNA | BC081934.1 | - | 6 - 7 | 2200 | 7114574 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |