Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_575106.5 | XP_575106.2 | 1 - 2 | 126 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |