Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025730.2 | NP_001020901.1 | 6 - 7 | 1232 | 54 | |
XM_006233623.2 | XP_006233685.1 | 5 - 6 | 1591 | 54 | ||
XM_006233624.2 | XP_006233686.1 | 7 - 8 | 1294 | 54 | ||
XM_006233625.2 | XP_006233687.1 | 7 - 8 | 1536 | 54 | ||
XM_006233626.2 | XP_006233688.1 | 6 - 7 | 1188 | 54 | ||
XM_006233627.2 | XP_006233689.1 | 6 - 7 | 1432 | 54 | ||
XM_006233628.2 | XP_006233690.1 | 5 - 6 | 948 | 54 | ||
GenBank mRNA | BC099179.1 | - | 6 - 7 | 1235 | 7383052 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |