Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246661.1 | NP_001233590.1 | 4 - 5 | 369 | 127 | |
GenBank mRNA | BC082799.1 | - | 4 - 5 | 369 | 7304460 | 127 |
FQ221000.1 | - | 4 - 5 | 419 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |