Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsb66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181694.2 | NP_859045.2 | 3 - 4 | 429 | 101 | |
XM_006233762.1 | XP_006233824.1 | 3 - 4 | 592 | 101 | ||
XM_006233763.2 | XP_006233825.1 | 3 - 4 | 808 | 101 | ||
XM_006233764.2 | XP_006233826.1 | 3 - 4 | 1263 | 101 | ||
XM_006233766.1 | XP_006233828.1 | 3 - 4 | 592 | 101 | ||
GenBank mRNA | BC097268.1 | - | 3 - 4 | 429 | 7457912 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |