Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033565.1 | NP_001028737.1 | 7 - 8 | 1167 | 115 | |
XM_006233639.2 | XP_006233701.1 | 8 - 9 | 1439 | 115 | ||
XM_006233640.2 | XP_006233702.1 | 9 - 10 | 1533 | 115 | ||
XM_006233642.2 | XP_006233704.1 | 8 - 9 | 1424 | 115 | ||
XM_006233643.2 | XP_006233705.1 | 8 - 9 | 1409 | 115 | ||
GenBank mRNA | AY536920.1 | - | 7 - 8 | 1167 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |