Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168549.1 | NP_001162021.1 | 10 - 11 | 1710 | 111 | |
XM_006233672.2 | XP_006233734.1 | 10 - 11 | 1678 | 111 | ||
XM_006233673.2 | XP_006233735.1 | 9 - 10 | 1645 | 111 | ||
XM_006233674.2 | XP_006233736.1 | 9 - 10 | 1645 | 111 | ||
XM_006233675.2 | XP_006233737.1 | 10 - 11 | 1678 | 111 | ||
XM_006233677.2 | XP_006233739.1 | 5 - 6 | 590 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |