Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109540.1 | NP_001103010.1 | 3 - 4 | 820 | 133 | |
XM_008763597.1 | XP_008761819.1 | 2 - 3 | 317 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |