Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238067.2 | XP_006238129.1 | 3 - 4 | 686 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |