Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100549.1 | NP_001094019.1 | 4 - 5 | 440 | 82 | |
XM_008767019.1 | XP_008765241.1 | 4 - 5 | 451 | 82 | ||
GenBank mRNA | BC088242.1 | - | 3 - 4 | 252 | 82 | |
BC158704.1 | - | 4 - 5 | 408 | 9030006 | 82 | |
CV106852.1 | - | 4 - 5 | 440 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |