Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080936.1 | NP_001074405.1 | 2 - 3 | 277 | 79 | |
XM_006245063.2 | XP_006245125.1 | 8 - 9 | 1308 | 79 | ||
XM_006245064.2 | XP_006245126.1 | 8 - 9 | 1085 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |