Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233581.2 | XP_006233643.1 | 1 - 2 | 572 | 56 | |
GenBank mRNA | FQ210613.1 | - | 1 - 2 | 147 | 56 | |
FQ212796.1 | - | 1 - 2 | 434 | 56 | ||
FQ218819.1 | - | 1 - 2 | 147 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |