Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023094.3 | NP_075582.3 | 2 - 3 | 133 | 89 | |
GenBank mRNA | AF264018.2 | - | 2 - 3 | 133 | 89 | |
AF469945.1 | - | 2 - 3 | 133 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |