Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031973.1 | NP_114179.1 | 5 - 6 | 686 | 60 | |
GenBank mRNA | AB038232.1 | - | 5 - 6 | 683 | 60 | |
AB048711.1 | - | 5 - 6 | 686 | 60 | ||
BC078783.1 | - | 5 - 6 | 659 | 7110490 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |