Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138897.1 | NP_620252.1 | 7 - 8 | 1098 | 87 | |
GenBank mRNA | D50671.1 | - | 7 - 8 | 1098 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |