Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015016.1 | NP_001015016.1 | 3 - 4 | 332 | 82 | |
GenBank mRNA | BC091196.1 | - | 3 - 4 | 332 | 7374499 | 82 |
FQ233803.1 | - | 3 - 4 | 344 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |