Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106637.1 | NP_001100107.1 | 548 | 62 | ||
GenBank mRNA | BC168988.1 | - | 498 | 8369626 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |