Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107933.1 | NP_001101403.1 | 6 - 7 | 1493 | 137 | |
XM_006238056.2 | XP_006238118.1 | 8 - 9 | 3096 | 137 | ||
GenBank mRNA | BC168982.1 | - | 8 - 9 | 1701 | 9030533 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |