Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034147.1 | NP_001029319.1 | 2 - 3 | 227 | 126 | |
GenBank mRNA | BC100091.1 | - | 2 - 3 | 227 | 7371990 | 126 |
FQ213073.1 | - | 2 - 3 | 273 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |