Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001056825.4 | XP_001056825.3 | 5 - 6 | 586 | 65 | |
XM_006253546.2 | XP_006253608.1 | 5 - 6 | 866 | 65 | ||
XM_006253547.2 | XP_006253609.1 | 5 - 6 | 867 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |