Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013137.2 | NP_001013155.1 | 3 - 4 | 480 | 65 | |
GenBank mRNA | BC101896.1 | - | 3 - 4 | 480 | 7932648 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |