Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cplx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053878.1 | NP_446330.1 | 2 - 3 | 324 | 69 | |
XM_006253596.2 | XP_006253658.1 | 2 - 3 | 360 | 69 | ||
XM_006253597.2 | XP_006253659.1 | 3 - 4 | 603 | 69 | ||
XM_008771482.1 | XP_008769704.1 | 3 - 4 | 603 | 69 | ||
XM_008771483.1 | XP_008769705.1 | 3 - 4 | 521 | 69 | ||
GenBank mRNA | D70816.1 | - | 2 - 3 | 324 | 69 | |
U35099.1 | - | 3 - 4 | 312 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |