Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107336.1 | NP_001100806.1 | 4 - 5 | 662 | 57 | |
XM_006253619.2 | XP_006253681.1 | 4 - 5 | 674 | 57 | ||
GenBank mRNA | BC166772.1 | - | 4 - 5 | 654 | 7934143 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |