Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109666.1 | NP_001103136.1 | 10 - 11 | 1519 | 85 | |
XM_006235929.2 | XP_006235991.1 | 11 - 12 | 1489 | 85 | ||
XM_006235930.2 | XP_006235992.1 | 11 - 12 | 1651 | 85 | ||
XM_006235931.2 | XP_006235993.1 | 11 - 12 | 1486 | 85 | ||
XM_006235932.2 | XP_006235994.1 | 11 - 12 | 1480 | 85 | ||
XM_006235933.2 | XP_006235995.1 | 11 - 12 | 1477 | 85 | ||
XM_006235934.2 | XP_006235996.1 | 10 - 11 | 1399 | 85 | ||
GenBank mRNA | BC129119.1 | - | 10 - 11 | 1386 | 7115823 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |