Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022395.2 | NP_071790.1 | 3 - 4 | 343 | 90 | |
GenBank mRNA | BC078826.1 | - | 3 - 4 | 338 | 7113892 | 90 |
D13907.1 | - | 3 - 4 | 321 | 90 | ||
L12965.1 | - | 3 - 4 | 344 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |