Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022515.1 | NP_071960.1 | 3 - 4 | 212 | 97 | |
GenBank mRNA | BC058473.1 | - | 3 - 4 | 237 | 6918141 | 97 |
FQ209984.1 | - | 3 - 4 | 231 | 97 | ||
FQ220432.1 | - | 3 - 4 | 241 | 97 | ||
FQ221142.1 | - | 3 - 4 | 230 | 97 | ||
FQ221221.1 | - | 3 - 4 | 237 | 97 | ||
FQ221486.1 | - | 3 - 4 | 233 | 97 | ||
FQ221683.1 | - | 3 - 4 | 224 | 97 | ||
FQ221688.1 | - | 3 - 4 | 233 | 97 | ||
FQ221810.1 | - | 3 - 4 | 233 | 97 | ||
FQ221946.1 | - | 3 - 4 | 233 | 97 | ||
FQ221988.1 | - | 3 - 4 | 237 | 97 | ||
FQ222823.1 | - | 3 - 4 | 233 | 97 | ||
FQ223282.1 | - | 3 - 4 | 233 | 97 | ||
FQ224199.1 | - | 3 - 4 | 233 | 97 | ||
FQ224575.1 | - | 3 - 4 | 237 | 97 | ||
FQ224676.1 | - | 3 - 4 | 233 | 97 | ||
X78443.1 | - | 3 - 4 | 212 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |