Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007602.1 | NP_001007603.1 | 1 - 2 | 113 | 99 | |
GenBank mRNA | AB008807.1 | - | 1 - 2 | 96 | 98 | |
BC079363.1 | - | 1 - 2 | 113 | 7109248 | 99 | |
FQ218105.1 | - | 1 - 2 | 88 | 99 | ||
FQ220742.1 | - | 1 - 2 | 55 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |