Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022265.2 | NP_071601.2 | 5 - 6 | 784 | 123 | |
XM_006231613.2 | XP_006231675.1 | 6 - 7 | 1065 | 123 | ||
XM_006231614.2 | XP_006231676.1 | 6 - 7 | 1117 | 123 | ||
XM_006231615.2 | XP_006231677.1 | 6 - 7 | 963 | 123 | ||
XM_006231616.2 | XP_006231678.1 | 6 - 7 | 940 | 123 | ||
GenBank mRNA | AF239739.2 | - | 5 - 6 | 784 | 123 | |
BC167751.1 | - | 6 - 7 | 972 | 9027822 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |