Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 3 - 4 | 316 | 131 | |
GenBank mRNA | D00512.1 | - | 3 - 4 | 248 | 131 | |
D13921.1 | - | 3 - 4 | 248 | 131 | ||
FM065829.1 | - | 3 - 4 | 316 | 131 | ||
FM071064.1 | - | 1 - 2 | 104 | 131 | ||
FQ217320.1 | - | 3 - 4 | 254 | 131 | ||
FQ228108.1 | - | 3 - 4 | 303 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |