Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110838.1 | NP_001104308.1 | 2 - 3 | 142 | 114 | |
XM_006244192.2 | XP_006244254.1 | 2 - 3 | 254 | 114 | ||
XM_006244193.2 | XP_006244255.1 | 2 - 3 | 363 | 114 | ||
XM_006244194.2 | XP_006244256.1 | 2 - 3 | 279 | 114 | ||
XM_006244195.2 | XP_006244257.1 | 2 - 3 | 247 | 114 | ||
XM_008766717.1 | XP_008764939.1 | 2 - 3 | 279 | 114 | ||
XM_008766722.1 | XP_008764944.1 | 2 - 3 | 253 | 114 | ||
XM_008766724.1 | XP_008764946.1 | 2 - 3 | 247 | 114 | ||
XM_008766725.1 | XP_008764947.1 | 2 - 3 | 251 | 114 | ||
GenBank mRNA | FQ210480.1 | - | 2 - 3 | 157 | 114 | |
FQ211645.1 | - | 2 - 3 | 227 | 114 | ||
FQ215070.1 | - | 2 - 3 | 157 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |